missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Oxocyclobutanecarboxylic Acid 97.0+%, TCI America™
$37.29 - $100.67
Chemical Identifiers
| CAS | 23761-23-1 |
|---|---|
| Molecular Formula | C5H6O3 |
| Molecular Weight (g/mol) | 114.10 |
| MDL Number | MFCD00100900 |
| InChI Key | IENOFRJPUPTEMI-UHFFFAOYSA-N |
| Synonym | 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h |
| PubChem CID | 4913358 |
| IUPAC Name | 3-oxocyclobutane-1-carboxylic acid |
| SMILES | OC(=O)C1CC(=O)C1 |
Chemical Identifiers
| 23761-23-1 | |
| 114.10 | |
| IENOFRJPUPTEMI-UHFFFAOYSA-N | |
| 4913358 | |
| OC(=O)C1CC(=O)C1 |
| C5H6O3 | |
| MFCD00100900 | |
| 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h | |
| 3-oxocyclobutane-1-carboxylic acid |
Specifications
| 23761-23-1 | |
| White-Yellow | |
| MFCD00100900 | |
| 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h | |
| OC(=O)C1CC(=O)C1 | |
| 114.10 | |
| 114.10 | |
| Crystalline Powder |
| 70°C | |
| C5H6O3 | |
| 1 g | |
| IENOFRJPUPTEMI-UHFFFAOYSA-N | |
| 3-oxocyclobutane-1-carboxylic acid | |
| 4913358 | |
| ≥97.0% (GC,T) | |
| 3-Oxocyclobutanecarboxylic Acid |
Safety and Handling
TSCA : No