missing translation for 'onlineSavingsMsg'
Learn More

3-Oxocyclobutanecarboxylic Acid 97.0+%, TCI America™

Catalog No. O03925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03925G 5 g
O03921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03925G Supplier TCI America Supplier No. O03925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23761-23-1
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.10
MDL Number MFCD00100900
InChI Key IENOFRJPUPTEMI-UHFFFAOYSA-N
Synonym 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h
PubChem CID 4913358
IUPAC Name 3-oxocyclobutane-1-carboxylic acid
SMILES OC(=O)C1CC(=O)C1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 5 g
Formula Weight 114.10
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxocyclobutanecarboxylic Acid