missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Oxocyclobutanecarboxylic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 114.10 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Oxocyclobutanecarboxylic Acid |
Safety and Handling
| TSCA | No |