Learn More
3-Oxo-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 98-78-2 |
|---|---|
| Molecular Formula | C6H8O3 |
| Molecular Weight (g/mol) | 128.13 |
| MDL Number | MFCD01320173 |
| InChI Key | RDSNBKRWKBMPOP-UHFFFAOYNA-N |
| Synonym | 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid |
| PubChem CID | 227798 |
| ChEBI | CHEBI:71213 |
| IUPAC Name | 3-oxocyclopentane-1-carboxylic acid |
| SMILES | OC(=O)C1CCC(=O)C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC426340010
|
Thermo Scientific Chemicals
426340010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC426340050
|
Thermo Scientific Chemicals
426340050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 98-78-2 | |
| 128.13 | |
| RDSNBKRWKBMPOP-UHFFFAOYNA-N | |
| 227798 | |
| 3-oxocyclopentane-1-carboxylic acid |
| C6H8O3 | |
| MFCD01320173 | |
| 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid | |
| CHEBI:71213 | |
| OC(=O)C1CCC(=O)C1 |
Specifications
| 98-78-2 | |
| 100.0 | |
| Glass bottle | |
| MFCD01320173 | |
| 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid | |
| OC(=O)C1CCC(=O)C1 | |
| 128.13 | |
| CHEBI:71213 | |
| 97% |
| 96.0 | |
| 97% | |
| C6H8O3 | |
| 1g | |
| RDSNBKRWKBMPOP-UHFFFAOYNA-N | |
| 3-oxocyclopentane-1-carboxylic acid | |
| 227798 | |
| 128.13 | |
| 3-Oxo-1-cyclopentanecarboxylic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only