missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. o047325g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O047325G 25 g
O04735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O047325G Supplier TCI America Supplier No. O047325G
Only null left

Chemical Identifiers

CAS 26272-83-3
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD08544401
InChI Key UMFWNFVHKAJOSE-UHFFFAOYSA-N
Synonym 3-oxetanyl tosylate, toluene-4-sulfonic acid oxetan-3-yl ester, oxetan-3-yl 4-methylbenzene-1-sulfonate, 3-tosyloxy oxetane, 3-tosyloxyoxetane, 3-oxetanyl p-toluenesulfonate, oxetan-3-yl-4-methylbenzenesulfonate, p-toluenesulfonic acid 3-oxetanyl ester, acmc-1cgaz, 3-oxetanol, 3-4-methylbenzenesulfonate
PubChem CID 13153907
IUPAC Name oxetan-3-yl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2COC2

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 372°C
Quantity 25 g
Formula Weight 228.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxetanyl p-Toluenesulfonate