missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5756906

 View more versions of this product

Catalog No. AAH5756906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Oxetanyl p-toluenesulfonate
1.34
179°C (354°F)
MFCD08544401
3-oxetanyl tosylate, toluene-4-sulfonic acid oxetan-3-yl ester, oxetan-3-yl 4-methylbenzene-1-sulfonate, 3-tosyloxy oxetane, 3-tosyloxyoxetane, 3-oxetanyl p-toluenesulfonate, oxetan-3-yl-4-methylbenzenesulfonate, p-toluenesulfonic acid 3-oxetanyl ester, acmc-1cgaz, 3-oxetanol, 3-4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OC2COC2
228.262
228.27
26272-83-3
372°C
C10H12O4S
5 g
UMFWNFVHKAJOSE-UHFFFAOYSA-N
oxetan-3-yl 4-methylbenzenesulfonate
13153907
96%

Chemical Identifiers

26272-83-3
228.262
UMFWNFVHKAJOSE-UHFFFAOYSA-N
13153907
CC1=CC=C(C=C1)S(=O)(=O)OC2COC2
C10H12O4S
MFCD08544401
3-oxetanyl tosylate, toluene-4-sulfonic acid oxetan-3-yl ester, oxetan-3-yl 4-methylbenzene-1-sulfonate, 3-tosyloxy oxetane, 3-tosyloxyoxetane, 3-oxetanyl p-toluenesulfonate, oxetan-3-yl-4-methylbenzenesulfonate, p-toluenesulfonic acid 3-oxetanyl ester, acmc-1cgaz, 3-oxetanol, 3-4-methylbenzenesulfonate
oxetan-3-yl 4-methylbenzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only