missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanone 97.0+%, TCI America™

Catalog No. p-7134350
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04671G 1 g
O04675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04671G Supplier TCI America Supplier No. O04671G
Only null left

Chemical Identifiers

CAS 6704-31-0
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD09263255
InChI Key ROADCYAOHVSOLQ-UHFFFAOYSA-N
Synonym 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
PubChem CID 15024254
IUPAC Name oxetan-3-one
SMILES C1C(=O)CO1

Specifications

Boiling Point 106°C
Quantity 1 g
UN Number 1993
Formula Weight 72.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Oxetanone
TSCA No
Recommended Storage Freezer