missing translation for 'onlineSavingsMsg'
Learn More

3-Oxabicyclo[3.1.0]hexane-2,4-dione 98.0+%, TCI America™

Catalog No. p-7134337
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04341G 1 g
O04345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04341G Supplier TCI America Supplier No. O04341G
Only null left

Chemical Identifiers

CAS 5617-74-3
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00126929
InChI Key ZRMYHUFDVLRYPN-UHFFFAOYSA-N
Synonym 3-oxabicyclo 3.1.0 hexane-2,4-dione, 1,2-cyclopropanedicarboxylic anhydride, oxabicyclohexanedione, pubchem18320, cyclopropane-1,2-dicarboxylic anhydride, 2,4-dioxo-3-oxabicyclo 3.1.0 hexane, 3-oxa-bicyclo 3.1.0 hexane-2,4-dione, 3-oxabicyclo 3.1.0-hexane-2,4-dione, 3-oxa-bicyclo 3.1.0-hexane-2,4-dione
PubChem CID 2734721
IUPAC Name 3-oxabicyclo[3.1.0]hexane-2,4-dione
SMILES C1C2C1C(=O)OC2=O

Specifications

Melting Point 60°C
Color White
Boiling Point 114°C
Quantity 1 g
Formula Weight 112.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2,4-dione