missing translation for 'onlineSavingsMsg'
Learn More

3-Octyne 99.0+%, TCI America™

Catalog No. O01265ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01265ML Supplier TCI America Supplier No. O01265ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15232-76-5
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00027276
InChI Key UDEISTCPVNLKRJ-UHFFFAOYSA-N
Synonym 3-octyne, butylethylacetylene, ethylbutylacetylene, 3-octyne, homopolymer, 1-ethyl-2-butylacetylene, acmc-209d7q, c4h9c.$.cc2h5, udeistcpvnlkrj-uhfffaoysa, 3-octyne,, 3h-indolium, 2-4-methoxyphenyl methylhydrazono methyl-1,3,3-trimethyl-, formate
PubChem CID 84849
IUPAC Name oct-3-yne
SMILES CCCCC#CCC

Specifications

Color Colorless
Boiling Point 133°C
Quantity 5 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Octyne
TSCA No
Recommended Storage Refrigerator