missing translation for 'onlineSavingsMsg'
Learn More

3-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog No. AC296140100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296140100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296140100 Supplier Thermo Scientific Chemicals Supplier No. 296140100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65016-62-8 | Molecular Formula: C12H20S | Molecular Weight: 196.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65016-62-8
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 106°C to 107°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H20S
Refractive Index 1.4870 to 1.492
Quantity 10 g
Specific Gravity 0.91
Molecular Weight (g/mol) 196.36
Formula Weight 196.36
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Octylthiophene, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
TSCA TSCA

RUO – Research Use Only