missing translation for 'onlineSavingsMsg'
Learn More

3-Octen-2-one, 97%

Catalog No. AAA1465209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1465209 10 g
AAA1465218 50 g
AAA1465230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1465209 Supplier Thermo Scientific Chemicals Supplier No. A1465209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1669-44-9 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1669-44-9
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00015565
InChI Key ZCFOBLITZWHNNC-VOTSOKGWSA-N
Synonym 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
PubChem CID 5363229
IUPAC Name (E)-oct-3-en-2-one
SMILES CCCCC=CC(=O)C

Specifications

Density 0.85
Boiling Point 60°C (16 mmHg)
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.448
Quantity 10 g
UN Number UN1224
Beilstein 1742230
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 3-Octen-2-one
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 216-793-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only