missing translation for 'onlineSavingsMsg'
Learn More

3-Octanone 98.0+%, TCI America™

Catalog No. p-7134196
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O012225ML 25 mL
O0122500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O012225ML Supplier TCI America Supplier No. O012225ML
Only null left

Chemical Identifiers

CAS 106-68-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009515
InChI Key RHLVCLIPMVJYKS-UHFFFAOYSA-N
Synonym 3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural
PubChem CID 246728
ChEBI CHEBI:80946
IUPAC Name octan-3-one
SMILES CCCCCC(=O)CC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 2271
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Octanone
EINECSNumber (2)-0544
RTECSNumber RH1485000
TSCA Yes