Learn More
3-Octanone, 98%
CAS: 106-68-3 | C8H16O | 128.215 g/mol
$134.88 - $134.88
Chemical Identifiers
| CAS | 106-68-3 |
|---|---|
| Molecular Formula | C8H16O |
| Molecular Weight (g/mol) | 128.215 |
| MDL Number | MFCD00009515 |
| InChI Key | RHLVCLIPMVJYKS-UHFFFAOYSA-N |
| Synonym | 3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural |
| PubChem CID | 246728 |
| ChEBI | CHEBI:80946 |
| IUPAC Name | octan-3-one |
| SMILES | CCCCCC(=O)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1211322
|
Thermo Scientific Chemicals
A1211322 |
100 g |
Each for $134.88
|
|
|||||
Description
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
Chemical Identifiers
| 106-68-3 | |
| 128.215 | |
| RHLVCLIPMVJYKS-UHFFFAOYSA-N | |
| 246728 | |
| octan-3-one |
| C8H16O | |
| MFCD00009515 | |
| 3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural | |
| CHEBI:80946 | |
| CCCCCC(=O)CC |
Specifications
| 106-68-3 | |
| 0.822 | |
| 51°C (123°F) | |
| 1.415 | |
| 100 g | |
| 1700021 | |
| 3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural | |
| RHLVCLIPMVJYKS-UHFFFAOYSA-N | |
| octan-3-one | |
| 246728 | |
| 128.22 | |
| 3-Octanone |
| -23°C | |
| 167°C to 168°C | |
| C8H16O | |
| MFCD00009515 | |
| UN1224 | |
| 14,6084 | |
| Insoluble in water. | |
| CCCCCC(=O)CC | |
| 128.215 | |
| CHEBI:80946 | |
| 98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H226-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 203-423-0
RTECSNumber : RH1485000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only