missing translation for 'onlineSavingsMsg'
Learn More

3-Octanol, 97%

Catalog No. AAA1448830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1448830 250 g
AAA1448818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1448830 Supplier Thermo Scientific Chemicals Supplier No. A1448830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-98-0 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 589-98-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00004590
InChI Key NMRPBPVERJPACX-UHFFFAOYNA-N
Synonym 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
PubChem CID 11527
ChEBI CHEBI:80945
IUPAC Name octan-3-ol
SMILES CCCCCC(O)CC

Specifications

Melting Point -45°C
Density 0.82
Boiling Point 175°C to 176°C
Flash Point 68°C (154°F)
Odor Pleasant
Refractive Index 1.427
Quantity 250 g
Beilstein 1719310
Formula Weight 130.23
Percent Purity 97%
Chemical Name or Material 3-Octanol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-667-4
RTECSNumber RH0855000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only