missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-O-Ethyl-L-ascorbic Acid 98.0+%, TCI America™
$78.29 - $268.43
Chemical Identifiers
| CAS | 86404-04-8 |
|---|---|
| Molecular Formula | C8H12O6 |
| Molecular Weight (g/mol) | 204.178 |
| MDL Number | MFCD09261382 |
| InChI Key | ZGSCRDSBTNQPMS-UJURSFKZSA-N |
| PubChem CID | 150736 |
| IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one |
| SMILES | CCOC1=C(C(=O)OC1C(CO)O)O |
Chemical Identifiers
| 86404-04-8 | |
| 204.178 | |
| ZGSCRDSBTNQPMS-UJURSFKZSA-N | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one |
| C8H12O6 | |
| MFCD09261382 | |
| 150736 | |
| CCOC1=C(C(=O)OC1C(CO)O)O |
Specifications
| 86404-04-8 | |
| White | |
| MFCD09261382 | |
| ZGSCRDSBTNQPMS-UJURSFKZSA-N | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one | |
| 150736 | |
| ≥98.0% (HPLC,T) | |
| 3-O-Ethyl-L-ascorbic Acid |
| 114°C | |
| C8H12O6 | |
| 5 g | |
| CCOC1=C(C(=O)OC1C(CO)O)O | |
| 204.178 | |
| 204.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator