missing translation for 'onlineSavingsMsg'
Learn More

3-O-Ethyl-L-ascorbic Acid 98.0+%, TCI America™

Catalog No. E09265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09265G 5 g
E092625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09265G Supplier TCI America Supplier No. E09265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86404-04-8
Molecular Formula C8H12O6
Molecular Weight (g/mol) 204.178
MDL Number MFCD09261382
InChI Key ZGSCRDSBTNQPMS-UJURSFKZSA-N
PubChem CID 150736
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one
SMILES CCOC1=C(C(=O)OC1C(CO)O)O

Specifications

Melting Point 114°C
Color White
Quantity 5 g
Formula Weight 204.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-O-Ethyl-L-ascorbic Acid
TSCA No
Recommended Storage Refrigerator