missing translation for 'onlineSavingsMsg'
Learn More

3-Nonenoic Acid 98.0+%, TCI America™

Supplier:  TCI America N03121ML

 View more versions of this product

Catalog No. N03121ML


Only null left

Description

Description

Specifications

Specifications

3-Nonenoic Acid
C9H16O2
1 mL
ZBPYTVBKHKUNHG-VOTSOKGWSA-N
(3E)-non-3-enoic acid
5282723
≥98.0% (T)
4124-88-3
MFCD00059000
3265
CCCCC\C=C\CC(O)=O
156.23
156.23
Liquid

Chemical Identifiers

4124-88-3
156.23
ZBPYTVBKHKUNHG-VOTSOKGWSA-N
(3E)-non-3-enoic acid
C9H16O2
MFCD00059000
5282723
CCCCC\C=C\CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions