missing translation for 'onlineSavingsMsg'
Learn More

3-Nonenoic Acid 98.0+%, TCI America™

Catalog No. N03125ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03125ML 5 mL
N03121ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03125ML Supplier TCI America Supplier No. N03125ML

Chemical Identifiers

CAS 4124-88-3
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00059000
InChI Key ZBPYTVBKHKUNHG-VOTSOKGWSA-N
PubChem CID 5282723
IUPAC Name (3E)-non-3-enoic acid
SMILES CCCCC\C=C\CC(O)=O

Specifications

Quantity 5 mL
UN Number 3265
Formula Weight 156.23
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Nonenoic Acid
EINECSNumber (2)-0609
TSCA Yes