missing translation for 'onlineSavingsMsg'
Learn More

3-Nonen-2-one 96.0+%, TCI America™

Catalog No. N059225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N059225ML Supplier TCI America Supplier No. N059225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14309-57-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00010241
InChI Key HDKLIZDXVUCLHQ-BQYQJAHWSA-N
Synonym 3-nonen-2-one, trans-3-nonen-2-one, e-non-3-en-2-one, e-3-nonen-2-one, 3-nonen-2-one, 3e, 3e-3-nonen-2-one, 3e-non-3-en-2-one, unii-fr0k39of16, 3e-nonen-2-one, 3-nonen-2-one fhfi
PubChem CID 5317045
IUPAC Name (3E)-non-3-en-2-one
SMILES CCCCC\C=C\C(C)=O

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 mL
Formula Weight 140.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nonen-2-one