missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitropyrazole 98.0+%, TCI America™
$100.67 - $337.40
Chemical Identifiers
| CAS | 26621-44-3 |
|---|---|
| Molecular Formula | C3H3N3O2 |
| Molecular Weight (g/mol) | 113.08 |
| MDL Number | MFCD00159621,MFCD00238787 |
| InChI Key | MZRUFMBFIKGOAL-UHFFFAOYSA-N |
| Synonym | 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole |
| PubChem CID | 123419 |
| IUPAC Name | 5-nitro-1H-pyrazole |
| SMILES | [O-][N+](=O)C1=CC=NN1 |
Chemical Identifiers
| 26621-44-3 | |
| 113.08 | |
| MZRUFMBFIKGOAL-UHFFFAOYSA-N | |
| 123419 | |
| [O-][N+](=O)C1=CC=NN1 |
| C3H3N3O2 | |
| MFCD00159621,MFCD00238787 | |
| 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole | |
| 5-nitro-1H-pyrazole |
Specifications
| 26621-44-3 | |
| White-Yellow | |
| MFCD00159621,MFCD00238787 | |
| 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole | |
| [O-][N+](=O)C1=CC=NN1 | |
| 113.08 | |
| 113.08 | |
| Crystalline Powder |
| 177°C | |
| C3H3N3O2 | |
| 1 g | |
| MZRUFMBFIKGOAL-UHFFFAOYSA-N | |
| 5-nitro-1H-pyrazole | |
| 123419 | |
| ≥98.0% (GC) | |
| 3-Nitropyrazole |
Safety and Handling
TSCA : No