missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Nitropropiophenone 98.0+%, TCI America™
$301.98 - $879.84
Chemical Identifiers
| CAS | 17408-16-1 |
|---|---|
| Molecular Formula | C9H9NO3 |
| Molecular Weight (g/mol) | 179.175 |
| MDL Number | MFCD00051516 |
| InChI Key | VSPOTMOYDHRALZ-UHFFFAOYSA-N |
| Synonym | 3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one |
| PubChem CID | 87096 |
| IUPAC Name | 1-(3-nitrophenyl)propan-1-one |
| SMILES | CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-] |
Chemical Identifiers
| 17408-16-1 | |
| 179.175 | |
| VSPOTMOYDHRALZ-UHFFFAOYSA-N | |
| 87096 | |
| CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-] |
| C9H9NO3 | |
| MFCD00051516 | |
| 3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one | |
| 1-(3-nitrophenyl)propan-1-one |
Specifications
| 17408-16-1 | |
| Yellow | |
| MFCD00051516 | |
| 3'-nitropropiophenone, 1-3-nitrophenyl propan-1-one, m-nitropropiophenone, 3-nitrophenyl ethyl ketone, 1-propanone, 1-3-nitrophenyl, m-nitropropio-phenone, 3;-nitropropiophenone, acmc-209e7u, 3-nitropropiophenone, 1-3-nitrophenyl propan-l-one | |
| CCC(=O)C1=CC(=CC=C1)[N+](=O)[O-] | |
| 179.175 | |
| 179.18 | |
| Crystalline Powder |
| 100°C | |
| C9H9NO3 | |
| 5 g | |
| VSPOTMOYDHRALZ-UHFFFAOYSA-N | |
| 1-(3-nitrophenyl)propan-1-one | |
| 87096 | |
| ≥98.0% (GC) | |
| 3′-Nitropropiophenone |
Safety and Handling
TSCA : No