missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic Hydrazide 97.0+%, TCI America™

$148.25 - $465.89

Chemical Identifiers

CAS 3682-15-3
Molecular Formula C8H5N3O4
Molecular Weight (g/mol) 207.15
MDL Number MFCD00006889
InChI Key YVOBBFQJMOGQOU-UHFFFAOYSA-N
Synonym 2,3-Dihydro-5-nitro-1,4-phthalazinedione, 3-Nitrophthalhydrazide
PubChem CID 3525223
IUPAC Name 5-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione
SMILES [O-][N+](=O)C1=C2C(=O)NNC(=O)C2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06225G
View Documents
TCI America
N06225G
5 g
Each for $148.25
Only null left
 
N062225G
View Documents
TCI America
N062225G
25 g
Each for $465.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3682-15-3
207.15
YVOBBFQJMOGQOU-UHFFFAOYSA-N
3525223
[O-][N+](=O)C1=C2C(=O)NNC(=O)C2=CC=C1
C8H5N3O4
MFCD00006889
2,3-Dihydro-5-nitro-1,4-phthalazinedione, 3-Nitrophthalhydrazide
5-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione

Specifications

3682-15-3
MFCD00006889
2,3-Dihydro-5-nitro-1,4-phthalazinedione, 3-Nitrophthalhydrazide
[O-][N+](=O)C1=C2C(=O)NNC(=O)C2=CC=C1
207.15
207.15
Crystalline Powder
C8H5N3O4
5 g
YVOBBFQJMOGQOU-UHFFFAOYSA-N
5-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione
3525223
≥97.0% (T)
3-Nitrophthalic Hydrazide
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'