missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoyl chloride, 98%

CAS: 121-90-4 | C7H4ClNO3 | 185.57 g/mol

$40.79 - $40.79

Chemical Identifiers

CAS 121-90-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.57
MDL Number MFCD00007247
InChI Key NXTNASSYJUXJDV-UHFFFAOYSA-N
Synonym m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
PubChem CID 8495
IUPAC Name 3-nitrobenzoyl chloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155840050
SDS View Documents
Thermo Scientific Chemicals
155840050
5 g Glass bottle
Each for $40.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-90-4
185.57
NXTNASSYJUXJDV-UHFFFAOYSA-N
8495
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl
C7H4ClNO3
MFCD00007247
m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
3-nitrobenzoyl chloride

Specifications

121-90-4
100.0
Brown
275.0°C to 278.0°C
Authentic
Glass bottle
O2NC6H4COCl
5 g
1.42
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl
185.57
185.57
Low Melting Solid
97.5
30.0°C to 35.0°C
1.4200g/mL
>112°C
98%
C7H4ClNO3
MFCD00007247
09, 381
m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
NXTNASSYJUXJDV-UHFFFAOYSA-N
3-nitrobenzoyl chloride
8495
98%
3-Nitrobenzoyl chloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-505-9

RTECSNumber : DM6646000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only