missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzhydrazide 98.0+%, TCI America™

Catalog No. N01475G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N01475G Supplier TCI America Supplier No. N01475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 618-94-0
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00017059
InChI Key NQEWXLVDAVTOHM-UHFFFAOYSA-N
Synonym 3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
PubChem CID 3291498
IUPAC Name 3-nitrobenzohydrazide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzhydrazide