missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzhydrazide, 98+%

CAS: 618-94-0 | C7H7N3O3 | 181.151 g/mol

$81.73 - $275.38

Chemical Identifiers

CAS 618-94-0
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00017059
InChI Key NQEWXLVDAVTOHM-UHFFFAOYSA-N
Synonym 3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
PubChem CID 3291498
IUPAC Name 3-nitrobenzohydrazide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2080006
View Documents
Thermo Scientific Chemicals
B2080006
5 g
Each for $81.73
Only null left
 
AAB2080014
View Documents
Thermo Scientific Chemicals
B2080014
25 g
Each for $275.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

618-94-0
181.151
NQEWXLVDAVTOHM-UHFFFAOYSA-N
3291498
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN
C7H7N3O3
MFCD00017059
3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
3-nitrobenzohydrazide

Specifications

618-94-0
C7H7N3O3
5 g
3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN
181.151
181.15
3-Nitrobenzhydrazide
152°C to 155°C
MFCD00017059
645512
NQEWXLVDAVTOHM-UHFFFAOYSA-N
3-nitrobenzohydrazide
3291498
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-572-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only