missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America N0143500G
Specifications
| 3-Nitrobenzenesulfonyl Chloride | |
| 62°C | |
| C6H4ClNO4S | |
| 500 g | |
| 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
| 221.611 | |
| 221.61 | |
| Crystalline Powder |
| 121-51-7 | |
| White-Yellow | |
| MFCD00007435 | |
| 3261 | |
| MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
| 3-nitrobenzenesulfonyl chloride | |
| 8476 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 121-51-7 | |
| 221.611 | |
| MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
| 8476 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
| C6H4ClNO4S | |
| MFCD00007435 | |
| 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
| 3-nitrobenzenesulfonyl chloride |
Safety and Handling
TSCA : Yes