missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. N014325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N014325G 25 g
N0143500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N014325G Supplier TCI America Supplier No. N014325G

Chemical Identifiers

CAS 121-51-7
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007435
InChI Key MWWNNNAOGWPTQY-UHFFFAOYSA-N
Synonym 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
PubChem CID 8476
IUPAC Name 3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 221.61
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonyl Chloride