Learn More
3-Nitrobenzenesulfonyl chloride, 98%
CAS: 121-51-7 | C6H4ClNO4S | 221.611 g/mol
Supplier: Thermo Scientific Chemicals A1767022
Description
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.
Solubility
Hydrolyzes slowly in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.
Specifications
| 3-Nitrobenzenesulfonyl chloride | |
| 59°C to 63°C | |
| C6H4ClNO4S | |
| 100 g | |
| 746542 | |
| 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
| MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
| 3-nitrobenzenesulfonyl chloride | |
| 8476 | |
| 98% |
| 121-51-7 | |
| >110°C (230°F) | |
| MFCD00007435 | |
| UN3261 | |
| Moisture sensitive | |
| Hydrolyzes slowly in water. | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
| 221.611 | |
| 221.62 |
Chemical Identifiers
| 121-51-7 | |
| 221.611 | |
| MWWNNNAOGWPTQY-UHFFFAOYSA-N | |
| 8476 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
| C6H4ClNO4S | |
| MFCD00007435 | |
| 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u | |
| 3-nitrobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 204-476-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only