missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonyl chloride, 98%

Catalog No. AAA1767014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1767014 25 g
AAA1767022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1767014 Supplier Thermo Scientific Chemicals Supplier No. A1767014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-51-7 | C6H4ClNO4S | 221.611 g/mol

It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.

Chemical Identifiers

CAS 121-51-7
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007435
InChI Key MWWNNNAOGWPTQY-UHFFFAOYSA-N
Synonym 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
PubChem CID 8476
IUPAC Name 3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 59°C to 63°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 746542
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes slowly in water.
Formula Weight 221.62
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-476-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only