missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
$233.00 - $788.49
Chemical Identifiers
| CAS | 98-47-5 |
|---|---|
| Molecular Formula | C6H5NO5S |
| Molecular Weight (g/mol) | 203.168 |
| MDL Number | MFCD00064383 |
| InChI Key | ONMOULMPIIOVTQ-UHFFFAOYSA-N |
| Synonym | 3-nitrobenzenesulfonic acid, m-nitrobenzenesulfonic acid, benzenesulfonic acid, 3-nitro, 3-nitrobenzenesulphonic acid, m-nitrobenzensulfonic acid, 3-nitrophenylsulfonic acid, kyselina 3-nitrobenzensulfonova, benzenesulfonic acid, m-nitro, ccris 3131, kyselina nitrobenzen-m-sulfonova |
| PubChem CID | 7387 |
| SMILES | C1=CC(=CC(=C1)S(=O)(=O)O)[N+](=O)[O-] |
Chemical Identifiers
| 98-47-5 | |
| 203.168 | |
| ONMOULMPIIOVTQ-UHFFFAOYSA-N | |
| 7387 |
| C6H5NO5S | |
| MFCD00064383 | |
| 3-nitrobenzenesulfonic acid, m-nitrobenzenesulfonic acid, benzenesulfonic acid, 3-nitro, 3-nitrobenzenesulphonic acid, m-nitrobenzensulfonic acid, 3-nitrophenylsulfonic acid, kyselina 3-nitrobenzensulfonova, benzenesulfonic acid, m-nitro, ccris 3131, kyselina nitrobenzen-m-sulfonova | |
| C1=CC(=CC(=C1)S(=O)(=O)O)[N+](=O)[O-] |
Specifications
| 98-47-5 | |
| C6H5NO5S | |
| 5 g | |
| 3-nitrobenzenesulfonic acid, m-nitrobenzenesulfonic acid, benzenesulfonic acid, 3-nitro, 3-nitrobenzenesulphonic acid, m-nitrobenzensulfonic acid, 3-nitrophenylsulfonic acid, kyselina 3-nitrobenzensulfonova, benzenesulfonic acid, m-nitro, ccris 3131, kyselina nitrobenzen-m-sulfonova | |
| C1=CC(=CC(=C1)S(=O)(=O)O)[N+](=O)[O-] | |
| 7387 | |
| ≥98.0% (HPLC,T) | |
| 3-Nitrobenzenesulfonic Acid Hydrate |
| White-Yellow | |
| MFCD00064383 | |
| 2305 | |
| ONMOULMPIIOVTQ-UHFFFAOYSA-N | |
| 203.168 | |
| 203.17 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2005
RTECSNumber : DB7190000
TSCA : Yes