missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzeneboronic acid, 98%

CAS: 13331-27-6 | C6H6BNO4 | 166.93 g/mol

Supplier:  Thermo Scientific Chemicals A1333606

 View more versions of this product

Catalog No. AAA1333606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Nitrobenzeneboronic acid
∼285°C (decomposition)
MFCD00007193
2938638
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
(3-nitrophenyl)boronic acid
1677
98%
13331-27-6
C6H6BNO4
5 g
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
166.93
166.93

Chemical Identifiers

13331-27-6
166.93
ZNRGSYUVFVNSAW-UHFFFAOYSA-N
1677
OB(O)C1=CC=CC(=C1)[N+]([O-])=O
C6H6BNO4
MFCD00007193
3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
(3-nitrophenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : CY8980000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only