missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzeneboronic acid, 98%

Catalog No. AAA1333603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333603 1 g
AAA1333606 5 g
AAA1333614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1333603 Supplier Thermo Scientific Chemicals Supplier No. A1333603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13331-27-6 | C6H6BNO4 | 166.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13331-27-6
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00007193
InChI Key ZNRGSYUVFVNSAW-UHFFFAOYSA-N
Synonym 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
PubChem CID 1677
IUPAC Name (3-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point ∼285°C (decomposition)
Quantity 1 g
Beilstein 2938638
Formula Weight 166.93
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber CY8980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only