missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzamidoxime, 97%, Thermo Scientific Chemicals

Catalog No. p-7034526
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5221206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5221206 Supplier Thermo Scientific Chemicals Supplier No. H5221206
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5023-94-9
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.15
MDL Number MFCD00053609
InChI Key ZAIHFKLUPWFUGH-UHFFFAOYSA-N
Synonym 3-nitrobenzamidoxime, n-hydroxy-3-nitro-benzamidine, m-nitrobenzamide oxime, z-n'-hydroxy-3-nitrobenzimidamide, m-nitrobenzamidoxime, n-hydroxy-3-nitro-benzenecarboximidamide, n'-hydroxy-3-nitrobenzene-1-carboximidamide, benzenecarboximidamide, n-hydroxy-3-nitro, z-n'-hydroxy-3-nitrobenzene-1-carboximidamide
PubChem CID 6424871
IUPAC Name N'-hydroxy-3-nitrobenzenecarboximidamide
SMILES N\C(=N/O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 181.15
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzamidoxime
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only