missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-L-phenylalanine, 95%

CAS: 19883-74-0 | C9H10N2O4 | 210.19 g/mol

Supplier:  Thermo Scientific Chemicals H6302906

Quantity 5 g

 View more versions of this product

Catalog No. AAH6302906

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19883-74-0
210.19
YTHDRUZHNYKZGF-UHFFFAOYNA-N
2761812
NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O
C9H10N2O4
MFCD01860661
3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602
2-amino-3-(3-nitrophenyl)propanoic acid

Specifications

3-Nitro-L-phenylalanine
White
MFCD01860661
YTHDRUZHNYKZGF-UHFFFAOYNA-N
2-amino-3-(3-nitrophenyl)propanoic acid
210.19
210.19
Powder
19883-74-0
C9H10N2O4
3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602
NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O
5 g
2761812
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions