missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitro-L-phenylalanine, 95%
CAS: 19883-74-0 | C9H10N2O4 | 210.19 g/mol
Supplier: Thermo Scientific Chemicals H6302903
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19883-74-0 | |
| 210.19 | |
| YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
| 2761812 | |
| NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O |
| C9H10N2O4 | |
| MFCD01860661 | |
| 3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602 | |
| 2-amino-3-(3-nitrophenyl)propanoic acid |
Specifications
| 3-Nitro-L-phenylalanine | |
| White | |
| MFCD01860661 | |
| YTHDRUZHNYKZGF-UHFFFAOYNA-N | |
| 2-amino-3-(3-nitrophenyl)propanoic acid | |
| 210.19 | |
| 210.19 | |
| Powder |
| 19883-74-0 | |
| C9H10N2O4 | |
| 3-nitro-l-phenylalanine, s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophenylalanine, 3-nitrophenylalanine, l-3-nitro phenylalanine, 2s-2-amino-3-3-nitrophenyl propanoic acid, l-3-nitrophe, melphalan impurity 2, l-3-no2-phe-oh, ncistruc1_000602 | |
| NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O | |
| 1 g | |
| 2761812 | |
| 95% |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C