missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-D-phenylalanine, 95%

Catalog No. AAH6348303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6348303 1 g
AAH63483MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6348303 Supplier Thermo Scientific Chemicals Supplier No. H6348303
Only null left

CAS: 169530-97-6 | C9H10N2O4 | 210.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 169530-97-6
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
MDL Number MFCD04117829
InChI Key YTHDRUZHNYKZGF-UHFFFAOYNA-N
Synonym r-2-amino-3-3-nitrophenyl propanoic acid, 3-nitro-d-phenylalanine, d-3-nitrophenylalanine, 2r-2-amino-3-3-nitrophenyl propanoic acid, d-3-no2-phe-oh, d-3-nitrophe, d-3-nitro phenylalanine, r-2-amino-3-3-nitrophenyl propanoicacid
PubChem CID 7000156
IUPAC Name 2-amino-3-(3-nitrophenyl)propanoic acid
SMILES NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O

Specifications

Color White to Yellow
Quantity 1 g
Formula Weight 210.19
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Nitro-D-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only