missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1,2,4-triazole 98.0+%, TCI America™

Catalog No. N04771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04771G 1 g
N04775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04771G Supplier TCI America Supplier No. N04771G

Chemical Identifiers

CAS 24807-55-4
Molecular Formula C2H2N4O2
Molecular Weight (g/mol) 114.064
MDL Number MFCD00009749
InChI Key KUEFXPHXHHANKS-UHFFFAOYSA-N
Synonym 3-nitro-1,2,4-triazole, 3-nitro-1h-1,2,4-triazole, 3-nitro-4h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro, s-triazole, 3-nitro, 5-nitro-1h-1,2,4 triazole, 3-nitro-1,2,4 triazole, zlchem 554, 3-nitro-1,4-triazole
PubChem CID 90614
IUPAC Name 5-nitro-1H-1,2,4-triazole
SMILES C1=NNC(=N1)[N+](=O)[O-]

Specifications

Melting Point 214°C
Color Yellow
Quantity 1 g
Formula Weight 114.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitro-1,2,4-triazole [Coupling Agent]
TSCA No
Recommended Storage Refrigerator