missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-n-Octylthiophene 98.0+%, TCI America™
$44.28 - $571.77
Chemical Identifiers
| CAS | 65016-62-8 |
|---|---|
| Molecular Formula | C12H20S |
| Molecular Weight (g/mol) | 196.352 |
| MDL Number | MFCD00085281 |
| InChI Key | WQYWXQCOYRZFAV-UHFFFAOYSA-N |
| Synonym | 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691 |
| PubChem CID | 566852 |
| IUPAC Name | 3-octylthiophene |
| SMILES | CCCCCCCCC1=CSC=C1 |
Chemical Identifiers
| 65016-62-8 | |
| 196.352 | |
| WQYWXQCOYRZFAV-UHFFFAOYSA-N | |
| 566852 | |
| CCCCCCCCC1=CSC=C1 |
| C12H20S | |
| MFCD00085281 | |
| 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691 | |
| 3-octylthiophene |
Specifications
| 65016-62-8 | |
| 106°C | |
| MFCD00085281 | |
| 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691 | |
| CCCCCCCCC1=CSC=C1 | |
| 196.352 | |
| 196.35 | |
| Liquid |
| Colorless | |
| C12H20S | |
| 1 g | |
| WQYWXQCOYRZFAV-UHFFFAOYSA-N | |
| 3-octylthiophene | |
| 566852 | |
| ≥98.0% (GC) | |
| 3-n-Octylthiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator