missing translation for 'onlineSavingsMsg'
Learn More

3-n-Octylthiophene 98.0+%, TCI America™

Catalog No. O02131G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02131G 1 g
O02135G 5 g
O021325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O02131G Supplier TCI America Supplier No. O02131G

Chemical Identifiers

CAS 65016-62-8
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

Color Colorless
Boiling Point 106°C
Quantity 1 g
Formula Weight 196.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-n-Octylthiophene
TSCA No
Recommended Storage Refrigerator