missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 322.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |