missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(N-Acetyl-N-ethylamino)pyrrolidine 98.0+%, TCI America™
Supplier: TCI America A11135G
Specifications
| 3-(N-Acetyl-N-ethylamino)pyrrolidine | |
| Yellow | |
| MFCD00059021 | |
| NHMWGGURJSUYGU-UHFFFAOYSA-N | |
| N-ethyl-N-pyrrolidin-3-ylacetamide | |
| 2756274 | |
| ≥98.0% (T) |
| 115445-29-9 | |
| C8H16N2O | |
| 5 g | |
| CCN(C1CCNC1)C(=O)C | |
| 156.229 | |
| 156.23 | |
| Liquid |
Chemical Identifiers
| 115445-29-9 | |
| 156.229 | |
| NHMWGGURJSUYGU-UHFFFAOYSA-N | |
| N-ethyl-N-pyrrolidin-3-ylacetamide |
| C8H16N2O | |
| MFCD00059021 | |
| 2756274 | |
| CCN(C1CCNC1)C(=O)C |
Safety and Handling
TSCA : No