missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholinomethyl)benzaldehyde, 95%, Thermo Scientific™

Catalog No. p-4510611
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC40104CB 250mg
CC40104DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC40104CB Supplier Thermo Scientific Chemicals Supplier No. CC40104CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446866-83-7
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
InChI Key KMYNYFKOOXFQGB-UHFFFAOYSA-N
Synonym 3-morpholinomethyl benzaldehyde, 3-morpholin-4-ylmethyl benzaldehyde, benzaldehyde,3-4-morpholinylmethyl, 3-morpholin-4-ylmethyl-benzaldehyde, 3-4-morpholinylmethyl benzaldehyde, 3-morpholinomethylbenzaldehyde, 3-morpholin-4-yl methyl benzaldehyde
PubChem CID 7164603
IUPAC Name 3-(morpholin-4-ylmethyl)benzaldehyde
SMILES C1COCCN1CC2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 250mg
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 3-(Morpholinomethyl)benzaldehyde