missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Morpholinomethyl)benzaldehyde, 95%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Quantity | 250mg |
| Formula Weight | 205.26 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-(Morpholinomethyl)benzaldehyde |