missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate, 97%, Thermo Scientific™

Catalog No. CC40151CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC40151CB 250mg
CC40151DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC40151CB Supplier Thermo Scientific Chemicals Supplier No. CC40151CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137605-80-2
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD09064949
InChI Key UOMMCPQFAXOMMO-UHFFFAOYSA-N
Synonym 3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 24229486
IUPAC Name 3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride
SMILES C1COCCN1CC2=CC=CC(=C2)C(=O)O.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 275.74
Percent Purity 97%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing