missing translation for 'onlineSavingsMsg'
Learn More

3-Methylvaleric Acid 98.0+%, TCI America™

Catalog No. M056910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M056910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M056910G Supplier TCI America Supplier No. M056910G

Chemical Identifiers

CAS 105-43-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002729,MFCD02262177
InChI Key IGIDLTISMCAULB-UHFFFAOYNA-N
Synonym 3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid
PubChem CID 7755
IUPAC Name 3-methylpentanoic acid
SMILES CCC(C)CC(O)=O

Specifications

Color Colorless
Boiling Point 197°C
Quantity 10 g
UN Number 3265
Formula Weight 116.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylvaleric Acid
EINECSNumber (2)-0608
TSCA Yes