Learn More
3-Methylvaleric acid, 99%
CAS: 105-43-1 | C6H12O2 | 116.16 g/mol
Supplier: Thermo Scientific Chemicals A1888614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Methylvaleric acid | |
| -42°C | |
| 196°C to 198°C | |
| Unpleasant | |
| 1.416 | |
| 25 g | |
| 1720696 | |
| IGIDLTISMCAULB-UHFFFAOYNA-N | |
| 3-methylpentanoic acid | |
| 7755 | |
| 99% |
| 105-43-1 | |
| 0.93 | |
| 85°C (185°F) | |
| C6H12O2 | |
| MFCD00002729,MFCD02262177 | |
| UN3265 | |
| 3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid | |
| CCC(C)CC(O)=O | |
| 116.16 | |
| 116.16 |
Chemical Identifiers
| 105-43-1 | |
| 116.16 | |
| IGIDLTISMCAULB-UHFFFAOYNA-N | |
| 7755 | |
| CCC(C)CC(O)=O |
| C6H12O2 | |
| MFCD00002729,MFCD02262177 | |
| 3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid | |
| 3-methylpentanoic acid |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 203-297-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only