missing translation for 'onlineSavingsMsg'
Learn More

3-Methylvaleric acid, 99%

Catalog No. AAA1888622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1888622 100 g
AAA1888606 5 g
AAA1888614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1888622 Supplier Thermo Scientific Chemicals Supplier No. A1888622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-43-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-43-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002729,MFCD02262177
InChI Key IGIDLTISMCAULB-UHFFFAOYNA-N
Synonym 3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid
PubChem CID 7755
IUPAC Name 3-methylpentanoic acid
SMILES CCC(C)CC(O)=O

Specifications

Melting Point -42°C
Density 0.93
Boiling Point 196°C to 198°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.416
Quantity 100 g
UN Number UN3265
Beilstein 1720696
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 3-Methylvaleric acid
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-297-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only