missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylthiophene 98.0+%, TCI America™
$61.51 - $725.11
Chemical Identifiers
| CAS | 616-44-4 |
|---|---|
| Molecular Formula | C5H6S |
| Molecular Weight (g/mol) | 98.163 |
| MDL Number | MFCD00005470 |
| InChI Key | QENGPZGAWFQWCZ-UHFFFAOYSA-N |
| Synonym | thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer |
| PubChem CID | 12024 |
| IUPAC Name | 3-methylthiophene |
| SMILES | CC1=CSC=C1 |
Chemical Identifiers
| 616-44-4 | |
| 98.163 | |
| QENGPZGAWFQWCZ-UHFFFAOYSA-N | |
| 12024 | |
| CC1=CSC=C1 |
| C5H6S | |
| MFCD00005470 | |
| thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer | |
| 3-methylthiophene |
Specifications
| 616-44-4 | |
| 115°C | |
| MFCD00005470 | |
| 1993 | |
| QENGPZGAWFQWCZ-UHFFFAOYSA-N | |
| 3-methylthiophene | |
| 12024 | |
| ≥98.0% (GC) | |
| 3-Methylthiophene |
| Yellow | |
| C5H6S | |
| 25 g | |
| thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer | |
| CC1=CSC=C1 | |
| 98.163 | |
| 98.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-0072
RTECSNumber : XM9800000
TSCA : Yes