missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene 98.0+%, TCI America™

Catalog No. M044025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044025G 25 g
M0440500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M044025G Supplier TCI America Supplier No. M044025G

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 1993
Formula Weight 98.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene
EINECSNumber (5)-0072
RTECSNumber XM9800000
TSCA Yes