missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 98+%

CAS: 616-44-4 | C5H6S | 98.163 g/mol

$210.07 - $925.01

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1339422
View Documents
Thermo Scientific Chemicals
A1339422
100 g
Each for $210.07
Only null left
 
AAA1339436
View Documents
Thermo Scientific Chemicals
A1339436
500 g
Each for $925.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-44-4
98.163
QENGPZGAWFQWCZ-UHFFFAOYSA-N
12024
CC1=CSC=C1
C5H6S
MFCD00005470
thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
3-methylthiophene

Specifications

616-44-4
1.02
11°C (51°F)
C5H6S
MFCD00005470
UN1993
thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
CC1=CSC=C1
98.163
98.17
3-Methylthiophene
-69°C
115°C to 117°C
Aromatic
1.52
100 g
1300
QENGPZGAWFQWCZ-UHFFFAOYSA-N
3-methylthiophene
12024
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H312-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 210-482-6

RTECSNumber : XM9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only